Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC684376 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229922.1 | XP_006229984.1 | 2 | 806 |
Gene Symbol: LOC684171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748935.2 | XP_003748983.1 | 1 | 836 |