Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM131B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031690.2 | NP_001026860.2 | 2, 3 | 287 | ||
NM_001278297.1 | NP_001265226.1 | 2, 3 | 287 | |||
NM_014690.4 | NP_055505.3 | 2, 3 | 180 | |||
XM_005250073.3 | XP_005250130.1 | 2, 3 | 501 | |||
XM_006716186.2 | XP_006716249.1 | 2, 3 | 180 | |||
GenBank mRNA | AK124557.1 | - | 2, 3 | 235 | ||
AK291470.1 | - | 2, 3 | 152 | |||
AL832579.2 | - | 1, 2 | 1768 | |||
BC045611.1 | - | 2, 3 | 292 | 5300056 | ||
BC050543.1 | - | 2, 3 | 155 | 5768309 | ||
DA226407.1 | - | 2, 3 | 180 |