Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100912233 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250508.1 | XP_006250570.1 | 2 | 346 |
Gene Symbol: LOC100910292 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250720.1 | XP_006250782.1 | 2 | 343 |
Gene Symbol: RGD1562423 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250712.1 | XP_006250774.1 | 2 | 349 |