Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294335.1 | NP_001281264.1 | 14 | 2526 | ||
NM_001294336.1 | NP_001281265.1 | 14 | 2526 | |||
XM_011541848.2 | XP_011540150.1 | 13 | 2480 | |||
XM_011541849.2 | XP_011540151.1 | 12 | 2315 | |||
XM_011541858.2 | XP_011540160.1 | 11 | 1479 | |||
XM_017001899.1 | XP_016857388.1 | 13 | 2610 | |||
XM_017001900.1 | XP_016857389.1 | 12 | 2582 | |||
XM_017001901.1 | XP_016857390.1 | 12 | 2582 | |||
XM_017001902.1 | XP_016857391.1 | 12 | 2583 | |||
XM_017001903.1 | XP_016857392.1 | 11 | 2514 | |||
XM_017001904.1 | XP_016857393.1 | 12 | 2579 | |||
XM_017001905.1 | XP_016857394.1 | 12 | 2581 | |||
XM_017001906.1 | XP_016857395.1 | 12 | 2580 | |||
XM_017001907.1 | XP_016857396.1 | 11 | 2414 | |||
XM_017001908.1 | XP_016857397.1 | 11 | 2415 | |||
XM_017001909.1 | XP_016857398.1 | 11 | 2416 | |||
XM_017001910.1 | XP_016857399.1 | 11 | 2414 | |||
XM_017001911.1 | XP_016857400.1 | 11 | 2415 | |||
XM_017001912.1 | XP_016857401.1 | 10 | 2248 | |||
GenBank mRNA | AB005298.1 | - | 14 | 2498 | ||
AK294999.1 | - | 7 | 1048 | |||
AK296502.1 | - | 9 | 1158 | |||
BC136533.1 | - | 12 | 2194 | 9020523 |
Set Membership: |
G-protein coupled receptor |