Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145320.1 | NP_001138792.1 | 6 | 1067 | ||
NM_014694.3 | NP_055509.2 | 6 | 725 | |||
XM_005272237.3 | XP_005272294.1 | 6 | 922 | |||
XM_005272238.3 | XP_005272295.1 | 5 | 569 | |||
XM_005272239.2 | XP_005272296.1 | 6 | 600 | |||
XM_011519241.2 | XP_011517543.2 | 5 | 748 | |||
XM_011519242.2 | XP_011517544.1 | 5 | 599 | |||
GenBank mRNA | AB011177.1 | - | 6 | 1067 | ||
BC050544.1 | - | 6 | 725 | 6196393 |