Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pin4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752069.3 | XP_003752117.1 | 3 | 276 | ||
XM_006257122.2 | XP_006257184.1 | 3 | 319 | |||
GenBank mRNA | FQ222828.1 | - | 3 | 219 |