Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTP14C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021645.5 | NP_067677.4 | 2 | 2950 | ||
GenBank mRNA | BC089407.1 | - | 1 | 2359 | 3065365 | |
BC122536.1 | - | 2 | 2727 | 40036054 | ||
D87455.1 | - | 2 | 2950 |