Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122776.1 | NP_001116248.1 | 2 | 1159 | ||
GenBank mRNA | X62841.1 | - | 2 | 1159 |