Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM63A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014698.2 | NP_055513.2 | 8 | 1166 | ||
XM_011544328.2 | XP_011542630.1 | 8 | 1018 | |||
XM_011544329.2 | XP_011542631.1 | 8 | 2285 | |||
XM_011544330.2 | XP_011542632.1 | 7 | 975 | |||
XM_011544331.2 | XP_011542633.1 | 8 | 849 | |||
XM_017002916.1 | XP_016858405.1 | 8 | 820 | |||
GenBank mRNA | AB018335.1 | - | 8 | 794 | ||
AK022502.1 | - | 8 | 815 | |||
AK222934.1 | - | 8 | 796 | |||
AK222944.1 | - | 8 | 796 | |||
AK309210.1 | - | 6 | 743 | |||
BC030245.1 | - | 8 | 799 | 5216414 |