Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294335.1 | NP_001281264.1 | 22 | 3550 | ||
NM_001294336.1 | NP_001281265.1 | 22 | 3550 | |||
XM_011541848.2 | XP_011540150.1 | 21 | 3504 | |||
XM_011541849.2 | XP_011540151.1 | 20 | 3339 | |||
XM_011541858.2 | XP_011540160.1 | 19 | 2503 | |||
XM_017001899.1 | XP_016857388.1 | 21 | 3634 | |||
XM_017001900.1 | XP_016857389.1 | 20 | 3606 | |||
XM_017001901.1 | XP_016857390.1 | 20 | 3606 | |||
XM_017001902.1 | XP_016857391.1 | 20 | 3607 | |||
XM_017001903.1 | XP_016857392.1 | 19 | 3538 | |||
XM_017001904.1 | XP_016857393.1 | 20 | 3603 | |||
XM_017001905.1 | XP_016857394.1 | 20 | 3605 | |||
XM_017001906.1 | XP_016857395.1 | 20 | 3604 | |||
XM_017001907.1 | XP_016857396.1 | 19 | 3438 | |||
XM_017001908.1 | XP_016857397.1 | 19 | 3439 | |||
XM_017001909.1 | XP_016857398.1 | 19 | 3440 | |||
XM_017001910.1 | XP_016857399.1 | 19 | 3438 | |||
XM_017001911.1 | XP_016857400.1 | 19 | 3439 | |||
XM_017001912.1 | XP_016857401.1 | 18 | 3272 | |||
GenBank mRNA | AB005298.1 | - | 22 | 3522 | ||
AK123392.1 | - | 1 | 339 | |||
AK294361.1 | - | 2 | 159 | |||
AK296502.1 | - | 17 | 2182 | |||
BC136533.1 | - | 20 | 3218 | 9020523 |
Set Membership: |
G-protein coupled receptor |