Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC501456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771014.1 | XP_008769236.1 | 1 | 54 |
Gene Symbol: LOC685976 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766820.1 | XP_008765042.1 | 7 | 1907 |