Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SECISBP2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193489.1 | NP_001180418.1 | 16 | 2627 | ||
NM_014701.3 | NP_055516.2 | 15 | 2492 | |||
GenBank mRNA | AK307480.1 | - | 16 | 2310 | ||
BC033001.2 | - | 16 | 2562 | 4825784 | ||
D87445.3 | - | 15 | 2362 |