Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamtor2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106441.1 | NP_001099911.1 | 2 | 252 | ||
XM_008761134.1 | XP_008759356.1 | 2 | 278 |