Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igsf21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271454.1 | NP_001258383.1 | 5 | 1120 | ||
XM_008764231.1 | XP_008762453.1 | 4 | 434 |