Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750667.3 | XP_003750715.3 | 2 | 397 | ||
XM_003750668.3 | XP_003750716.3 | 2 | 398 |