Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETD1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014712.2 | NP_055527.1 | 4, 5 | 1211 | ||
XM_005255723.1 | XP_005255780.1 | 4, 5 | 695 | |||
XM_006721106.3 | XP_006721169.1 | 4, 5 | 616 | |||
XM_017023909.1 | XP_016879398.1 | 4, 5 | 1848 | |||
GenBank mRNA | AB002337.2 | - | 4, 5 | 1211 | ||
BC035795.2 | - | 4, 5 | 711 |