Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAPTM4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014713.4 | NP_055528.1 | 5, 6 | 1044 | ||
GenBank mRNA | AK129875.1 | - | 5, 6 | 694 | ||
AY359028.1 | - | 5, 6 | 660 | |||
BC000421.2 | - | 5, 6 | 621 | 2964450 | ||
BC003158.1 | - | 5, 6 | 684 | 2958870 | ||
BC093075.1 | - | 5, 6 | 644 | 30402963 | ||
CR456758.1 | - | 5, 6 | 536 | |||
D14696.1 | - | 5, 6 | 682 |