Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014714.3 | NP_055529.2 | 11 | 1702 | ||
XM_005255725.4 | XP_005255782.1 | 10 | 1404 | |||
XM_005255726.3 | XP_005255783.1 | 10 | 1404 | |||
XM_006720990.3 | XP_006721053.1 | 10 | 1620 | |||
XM_006720991.3 | XP_006721054.1 | 10 | 1810 | |||
XM_011522766.2 | XP_011521068.1 | 10 | 1404 | |||
XM_011522767.2 | XP_011521069.1 | 3 | 422 | |||
XM_011522769.2 | XP_011521071.1 | 10 | 1404 | |||
XM_011522771.2 | XP_011521073.1 | 10 | 1404 | |||
XM_011522772.2 | XP_011521074.1 | 10 | 1404 | |||
XM_017023910.1 | XP_016879399.1 | 10 | 1393 | |||
XM_017023911.1 | XP_016879400.1 | 9 | 1371 | |||
GenBank mRNA | AB011162.1 | - | 10 | 1399 | ||
AK001797.1 | - | 261 | ||||
AK299035.1 | - | 10 | 1401 | |||
AK299115.1 | - | 5 | 576 | |||
BC035577.1 | - | 9 | 1387 | 5552740 |