Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142685.1 | NP_001136157.1 | 21 | 3293 | ||
NM_014715.3 | NP_055530.2 | 12 | 2625 | |||
XM_005271736.4 | XP_005271793.2 | 22 | 4323 | |||
XM_011543072.2 | XP_011541374.1 | 22 | 3525 | |||
XM_011543073.2 | XP_011541375.2 | 22 | 3261 | |||
XM_011543075.2 | XP_011541377.1 | 13 | 2471 | |||
XM_017018595.1 | XP_016874084.1 | 22 | 3242 | |||
XM_017018596.1 | XP_016874085.1 | 20 | 3519 | |||
GenBank mRNA | AB018255.2 | - | 12 | 2546 | ||
AB088416.1 | - | 12 | 2546 | |||
AL833062.2 | - | 19 | 3788 | |||
AY194287.1 | - | 12 | 2549 | |||
BC104898.1 | - | 11 | 2280 | 8143901 | ||
BC113429.1 | - | 11 | 2280 | 8322481 | ||
EF127492.1 | - | 21 | 3293 |