Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF536 SIRNA Assays ›
Gene Symbol:
ZNF536
Gene Name:
zinc finger protein 536
Gene Aliases:
-
Chromosome Location:
Chr.19: 30224236 - 30713538 on Build GRCh38
UniGene ID:
Hs.378901
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s18805
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF536
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014717.2
NP_055532.1
4
3513
XM_011527554.2
XP_011525856.1
7
5058
XM_011527555.2
XP_011525857.1
8
5147
XM_011527557.2
XP_011525859.1
5
4139
XM_011527558.2
XP_011525860.1
5
3938
XM_011527560.2
XP_011525862.1
4
4051
XM_017027527.1
XP_016883016.1
7
4125
XM_017027528.1
XP_016883017.1
6
4008
XM_017027529.1
XP_016883018.1
8
4214
XM_017027530.1
XP_016883019.1
7
3717
XM_017027531.1
XP_016883020.1
7
3812
XM_017027532.1
XP_016883021.1
7
3707
XM_017027533.1
XP_016883022.1
7
3682
XM_017027534.1
XP_016883023.1
8
4426
XM_017027535.1
XP_016883024.1
6
3600
XM_017027536.1
XP_016883025.1
6
4055
XM_017027537.1
XP_016883026.1
6
4038
XM_017027538.1
XP_016883027.1
7
4125
XM_017027539.1
XP_016883028.1
7
4125
XM_017027540.1
XP_016883029.1
6
4037
XM_017027541.1
XP_016883030.1
4
4050
XM_017027542.1
XP_016883031.1
6
4037
XM_017027543.1
XP_016883032.1
4
1907
GenBank mRNA
AB002388.1
-
4
3485
BC132720.1
-
4
3369
40146742
BC132722.1
-
4
3369
40146744
BC146757.1
-
4
3485
9007195
BC150171.1
-
4
3494
40147081
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
signal transduction
multicellular organism development
negative regulation of neuron differentiation
negative regulation of retinoic acid receptor signaling pathway
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
retinoic acid-responsive element binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-7745c474b-dptlh:80/100.66.78.226:80.
git-commit: c3c9db9cc82507548abdd0ae6a4ade3f81f66292
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.20.0-2024.11.45-1.0