Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206938.1 | NP_001193867.1 | 7 | 2661 | ||
NM_001206941.1 | NP_001193870.1 | 6 | 2032 | |||
NM_014719.2 | NP_055534.1 | 7 | 2621 | |||
XM_005250074.3 | XP_005250131.1 | 7 | 2661 | |||
XM_005250075.2 | XP_005250132.1 | 7 | 2663 | |||
XM_005250076.3 | XP_005250133.1 | 7 | 2698 | |||
XM_005250077.2 | XP_005250134.1 | 8 | 2827 | |||
XM_006716191.2 | XP_006716254.1 | 7 | 2627 | |||
XM_006716192.3 | XP_006716255.1 | 7 | 2770 | |||
XM_017012833.1 | XP_016868322.1 | 7 | 2693 | |||
XM_017012834.1 | XP_016868323.1 | 7 | 2663 | |||
XM_017012835.1 | XP_016868324.1 | 8 | 2827 | |||
XM_017012836.1 | XP_016868325.1 | 7 | 2627 | |||
GenBank mRNA | AB018281.1 | - | 7 | 2621 | ||
AK001640.1 | - | 7 | 2697 | |||
AK291605.1 | - | 7 | 2695 | |||
AK294312.1 | - | 3 | 641 | |||
AK299911.1 | - | 6 | 1930 | |||
BC000609.2 | - | 7 | 2602 | 3346754 |