Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Odf2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134708.1 | NP_001128180.1 | 7 | 773 | ||
XM_006233439.2 | XP_006233501.1 | 8 | 1213 | |||
XM_006233440.2 | XP_006233502.1 | 8 | 1357 | |||
XM_006233441.2 | XP_006233503.1 | 8 | 1341 | |||
XM_006233442.2 | XP_006233504.1 | 7 | 7201 | |||
XM_006233443.2 | XP_006233505.1 | 8 | 1345 | |||
XM_006233444.2 | XP_006233506.1 | 6 | 7121 | |||
XM_006233445.2 | XP_006233507.1 | 7 | 1265 | |||
XM_006233446.2 | XP_006233508.1 | 6 | 7068 | |||
XM_006233447.2 | XP_006233509.1 | 7 | 7195 | |||
XM_006233448.2 | XP_006233510.1 | 6 | 7120 | |||
XM_006233449.2 | XP_006233511.1 | 5 | 6991 | |||
XM_008761524.1 | XP_008759746.1 | 7 | 7197 | |||
XM_008761525.1 | XP_008759747.1 | 8 | 1485 | |||
GenBank mRNA | BC166823.1 | - | 7 | 773 | 8370140 | |
CB767200.1 | - | 3 | 240 |