Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Odf2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134708.1 | NP_001128180.1 | 12 | 1413 | ||
XM_006233439.2 | XP_006233501.1 | 13 | 1853 | |||
XM_006233440.2 | XP_006233502.1 | 13 | 1997 | |||
XM_006233441.2 | XP_006233503.1 | 13 | 1981 | |||
XM_006233442.2 | XP_006233504.1 | 12 | 7841 | |||
XM_006233443.2 | XP_006233505.1 | 13 | 1985 | |||
XM_006233444.2 | XP_006233506.1 | 11 | 7761 | |||
XM_006233445.2 | XP_006233507.1 | 12 | 1905 | |||
XM_006233446.2 | XP_006233508.1 | 11 | 7708 | |||
XM_006233447.2 | XP_006233509.1 | 12 | 7835 | |||
XM_006233448.2 | XP_006233510.1 | 11 | 7760 | |||
XM_006233449.2 | XP_006233511.1 | 10 | 7631 | |||
XM_008761524.1 | XP_008759746.1 | 12 | 7837 | |||
XM_008761525.1 | XP_008759747.1 | 13 | 2125 | |||
GenBank mRNA | BC166823.1 | - | 12 | 1547 | 8370140 |