Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100361898 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749083.3 | XP_003749131.1 | 1 | 560 |
Gene Symbol: Chchd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013431.1 | NP_001013449.1 | 3 | 643 | ||
GenBank mRNA | BC091407.1 | - | 3 | 643 | 7324948 | |
FQ220478.1 | - | 3 | 508 | |||
FQ220795.1 | - | 3 | 538 | |||
FQ224864.1 | - | 3 | 556 | |||
FQ229282.1 | - | 3 | 508 |