Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LZTS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282533.1 | NP_001269462.1 | 3 | 675 | ||
XM_005260890.3 | XP_005260947.1 | 3 | 619 | |||
XM_005260892.1 | XP_005260949.1 | 3 | 675 | |||
XM_005260893.2 | XP_005260950.1 | 3 | 588 | |||
XM_011529407.2 | XP_011527709.1 | 3 | 619 | |||
XM_011529408.2 | XP_011527710.1 | 3 | 1657 | |||
XM_011529409.2 | XP_011527711.1 | 3 | 2077 | |||
GenBank mRNA | AB011124.1 | - | 1 | 1703 | ||
BC038860.1 | - | 3 | 631 | 5260571 |