Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC685590 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773148.1 | XP_008771370.1 | 2 | 349 |
Gene Symbol: Pik3ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133399.2 | NP_596890.2 | 1 | 244 | ||
GenBank mRNA | CB793501.1 | - | 1 | 208 | ||
FM035395.1 | - | 1 | 54 |