Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105251.2 | NP_001098721.1 | 4 | 2456 | ||
NM_001284236.1 | NP_001271165.1 | 4 | 2382 | |||
NM_001284237.1 | NP_001271166.1 | 3 | 2328 | |||
NM_014733.4 | NP_055548.3 | 3 | 2328 | |||
XM_005248632.4 | XP_005248689.1 | 3 | 2454 | |||
XM_011543753.2 | XP_011542055.1 | 4 | 2353 | |||
XM_017010091.1 | XP_016865580.1 | 5 | 2514 | |||
XM_017010092.1 | XP_016865581.1 | 4 | 2500 | |||
XM_017010093.1 | XP_016865582.1 | 1 | 631 | |||
XM_017010094.1 | XP_016865583.1 | 1 | 631 | |||
XM_017010095.1 | XP_016865584.1 | 4 | 2353 | |||
GenBank mRNA | AB002303.1 | - | 4 | 2396 | ||
AF434817.1 | - | 2 | 2148 | |||
AL833087.2 | - | 1 | 1375 | |||
BC030808.1 | - | 3 | 2258 | 4701661 | ||
BX537424.1 | - | 3 | 2280 | |||
CR933621.1 | - | 4 | 2331 |