Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109477.2 | NP_001102947.2 | 2, 3 | 170 | ||
XM_006240352.2 | XP_006240414.1 | 3, 4 | 379 | |||
GenBank mRNA | BC161941.1 | - | 209 | |||
EV765393.1 | - | 3, 4 | 209 | |||
FQ211665.1 | - | 174 |