Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100363333 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250717.1 | XP_006250779.1 | 2 | 602 |
Gene Symbol: LOC685667 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250713.1 | XP_006250775.1 | 2 | 602 |