Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105251.2 | NP_001098721.1 | 12, 13 | 4009 | ||
NM_001284236.1 | NP_001271165.1 | 12, 13 | 3935 | |||
NM_014733.4 | NP_055548.3 | 11, 12 | 3881 | |||
XM_005248632.4 | XP_005248689.1 | 11, 12 | 4007 | |||
XM_011543753.2 | XP_011542055.1 | 12, 13 | 3906 | |||
XM_017010091.1 | XP_016865580.1 | 13, 14 | 4067 | |||
XM_017010092.1 | XP_016865581.1 | 12, 13 | 4053 | |||
XM_017010093.1 | XP_016865582.1 | 11, 12 | 2387 | |||
XM_017010094.1 | XP_016865583.1 | 10, 11 | 2332 | |||
XM_017010095.1 | XP_016865584.1 | 13, 14 | 3961 | |||
XM_017010096.1 | XP_016865585.1 | 8, 9 | 1409 | |||
GenBank mRNA | AB002303.1 | - | 12, 13 | 3949 | ||
AF434817.1 | - | 10, 11 | 3701 | |||
AK127003.1 | - | 7, 8 | 1098 | |||
AL833087.2 | - | 9, 10 | 2928 | |||
BX537424.1 | - | 11, 12 | 3833 | |||
BX649172.1 | - | 1 | 2483 | |||
CR933621.1 | - | 12, 13 | 3883 |