Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751642.2 | XP_003751690.1 | 2 | 266 | ||
GenBank mRNA | CO398682.1 | - | 77 |
Gene Symbol: LOC103694037 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771511.1 | XP_008769733.1 | 2 | 139 |