Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASSF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014737.2 | NP_055552.1 | 5 | 417 | ||
NM_170774.1 | NP_739580.1 | 4 | 275 | |||
XM_005260895.3 | XP_005260952.1 | 5 | 483 | |||
XM_011529410.1 | XP_011527712.1 | 5 | 411 | |||
XM_011529411.1 | XP_011527713.1 | 5 | 483 | |||
XM_011529412.2 | XP_011527714.1 | 4 | 332 | |||
XM_017028149.1 | XP_016883638.1 | 4 | 306 | |||
XM_017028150.1 | XP_016883639.1 | 6 | 620 | |||
XM_017028151.1 | XP_016883640.1 | 6 | 587 | |||
XM_017028152.1 | XP_016883641.1 | 6 | 664 | |||
XM_017028153.1 | XP_016883642.1 | 4 | 365 | |||
GenBank mRNA | AK222650.1 | - | 5 | 380 | ||
AK291458.1 | - | 5 | 372 | |||
AK307232.1 | - | 2 | 106 | |||
AY154470.1 | - | 5 | 417 | |||
AY154472.1 | - | 4 | 275 | |||
BC117118.1 | - | 3 | 254 | 40125669 | ||
BC117120.1 | - | 3 | 254 | 40125671 | ||
CR627436.1 | - | 5 | 437 | |||
D79990.1 | - | 5 | 417 | |||
DA333367.1 | - | 5 | 380 |