Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnpy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077585.1 | NP_001071053.1 | 5 | 807 | ||
GenBank mRNA | BC126101.1 | - | 5 | 807 | 7319422 | |
FQ220100.1 | - | 5 | 663 | |||
FQ220178.1 | - | 5 | 791 | |||
FQ225671.1 | - | 5 | 743 | |||
FQ231871.1 | - | 5 | 714 |