Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATG13 SIRNA Assays ›
Gene Symbol:
ATG13
Gene Name:
autophagy related 13
Gene Aliases:
KIAA0652, PARATARG8
Chromosome Location:
Chr.11: 46617276 - 46676019 on Build GRCh38
UniGene ID:
Hs.127403
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s18881
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATG13
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142673.2
NP_001136145.1
4, 5
841
NM_001205119.1
NP_001192048.1
3, 4
785
NM_001205120.1
NP_001192049.1
3, 4
785
NM_001205121.1
NP_001192050.1
4, 5
836
NM_001205122.1
NP_001192051.1
2, 3
703
NM_014741.4
NP_055556.2
4, 5
841
XM_005253262.2
XP_005253319.1
4, 5
581
XM_005253263.2
XP_005253320.1
4, 5
872
XM_005253265.2
XP_005253322.1
5, 6
1078
XM_005253266.2
XP_005253323.1
4, 5
807
XM_005253268.2
XP_005253325.1
2, 3
184
XM_006718394.2
XP_006718457.1
5, 6
928
XM_006718395.2
XP_006718458.1
5, 6
863
XM_006718396.2
XP_006718459.1
4, 5
962
XM_011520493.1
XP_011518795.1
6, 7
1065
XM_011520496.2
XP_011518798.1
3, 4
840
XM_011520499.2
XP_011518801.1
3, 4
454
XM_011520500.1
XP_011518802.1
5, 6
928
XM_011520501.1
XP_011518803.1
5, 6
928
XM_011520502.1
XP_011518804.1
5, 6
928
XM_011520503.1
XP_011518805.1
5, 6
928
XM_017018599.1
XP_016874088.1
5, 6
1013
XM_017018600.1
XP_016874089.1
4, 5
581
XM_017018601.1
XP_016874090.1
3, 4
756
XM_017018602.1
XP_016874091.1
5, 6
1078
XM_017018603.1
XP_016874092.1
6, 7
1195
XM_017018604.1
XP_016874093.1
4, 5
872
XM_017018605.1
XP_016874094.1
5, 6
863
XM_017018606.1
XP_016874095.1
4, 5
807
XM_017018607.1
XP_016874096.1
2, 3
184
XM_017018608.1
XP_016874097.1
4, 5
581
XM_017018609.1
XP_016874098.1
4, 5
872
XM_017018610.1
XP_016874099.1
3, 4
756
XM_017018611.1
XP_016874100.1
4, 5
807
XM_017018612.1
XP_016874101.1
5, 6
1078
XM_017018613.1
XP_016874102.1
4, 5
872
XM_017018614.1
XP_016874103.1
5, 6
863
XM_017018615.1
XP_016874104.1
4, 5
962
XM_017018616.1
XP_016874105.1
3, 4
756
XM_017018617.1
XP_016874106.1
5, 6
961
XM_017018618.1
XP_016874107.1
5, 6
928
XM_017018619.1
XP_016874108.1
4, 5
812
XM_017018620.1
XP_016874109.1
4, 5
872
XM_017018621.1
XP_016874110.1
3, 4
756
XM_017018622.1
XP_016874111.1
4, 5
824
GenBank mRNA
AB014552.1
-
3, 4
465
AK023867.1
-
4, 5
574
AK223603.1
-
3, 4
461
AK289641.1
-
4, 5
388
AK293109.1
-
4, 5
540
AK294110.1
-
2, 3
416
BC001331.1
-
4, 5
270
3458359
BC002378.2
-
4, 5
540
2961127
BC006191.2
-
4, 5
582
3936851
CD675643.1
-
2, 3
160
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
autophagosome assembly
macroautophagy
mitophagy in response to mitochondrial depolarization
positive regulation of protein targeting to mitochondrion
protein binding
protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-jlg4l:80/100.66.77.222:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline