Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM9SF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014742.3 | NP_055557.2 | 16 | 1898 | ||
XM_005260622.4 | XP_005260679.1 | 16 | 1950 | |||
XM_017028154.1 | XP_016883643.1 | 15 | 1595 | |||
XM_017028155.1 | XP_016883644.1 | 15 | 1866 | |||
XM_017028156.1 | XP_016883645.1 | 16 | 1646 | |||
XM_017028157.1 | XP_016883646.1 | 16 | 1903 | |||
XM_017028158.1 | XP_016883647.1 | 15 | 1557 | |||
XM_017028159.1 | XP_016883648.1 | 16 | 1703 | |||
XM_017028160.1 | XP_016883649.1 | 16 | 1615 | |||
XM_017028161.1 | XP_016883650.1 | 15 | 1567 | |||
GenBank mRNA | AK124411.1 | - | 1 | 940 | ||
AK126534.1 | - | 14 | 1348 | |||
AK294980.1 | - | 16 | 1646 | |||
AK296500.1 | - | 16 | 1555 | |||
AK316293.1 | - | 15 | 1602 | |||
BC021107.1 | - | 16 | 1674 | 4896167 | ||
BC022850.2 | - | 15 | 1656 | |||
D87444.1 | - | 16 | 1952 |