Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casq1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159594.1 | NP_001153066.1 | 4, 5 | 738 | ||
GenBank mRNA | CB699232.1 | - | 4, 5 | 325 | ||
FM056035.1 | - | 1, 2 | 43 | |||
FQ216689.1 | - | 4, 5 | 315 | |||
FQ224875.1 | - | 5, 6 | 565 |