Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM9SF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014742.3 | NP_055557.2 | 3 | 403 | ||
XM_005260622.4 | XP_005260679.1 | 3 | 455 | |||
XM_017028154.1 | XP_016883643.1 | 3 | 184 | |||
XM_017028155.1 | XP_016883644.1 | 3 | 455 | |||
XM_017028156.1 | XP_016883645.1 | 3 | 195 | |||
XM_017028157.1 | XP_016883646.1 | 3 | 452 | |||
XM_017028158.1 | XP_016883647.1 | 3 | 190 | |||
XM_017028159.1 | XP_016883648.1 | 3 | 236 | |||
XM_017028160.1 | XP_016883649.1 | 3 | 192 | |||
XM_017028161.1 | XP_016883650.1 | 3 | 184 | |||
GenBank mRNA | AK126534.1 | - | 3 | 195 | ||
AK294980.1 | - | 3 | 195 | |||
AK296500.1 | - | 3 | 183 | |||
AK302439.1 | - | 3 | 181 | |||
AK316293.1 | - | 2 | 107 | |||
BC021107.1 | - | 3 | 179 | 4896167 | ||
BC022850.2 | - | 2 | 161 | |||
D87444.1 | - | 3 | 457 | |||
DC381660.1 | - | 3 | 403 |