Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107337.1 | NP_001100807.1 | 23 | 6540 | ||
XM_006253620.2 | XP_006253682.1 | 23 | 6789 | |||
XM_006253621.2 | XP_006253683.1 | 23 | 6891 | |||
XM_006253622.2 | XP_006253684.1 | 23 | 6619 | |||
XM_006253623.2 | XP_006253685.1 | 23 | 6778 | |||
XM_006253624.2 | XP_006253686.1 | 23 | 6715 | |||
XM_006253625.2 | XP_006253687.1 | 23 | 6894 | |||
XM_008771491.1 | XP_008769713.1 | 23 | 6844 | |||
XM_008771492.1 | XP_008769714.1 | 22 | 5755 | |||
XM_008771493.1 | XP_008769715.1 | 22 | 6045 |