Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIEZO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142864.3 | NP_001136336.2 | 45 | 6764 | ||
GenBank mRNA | AB161230.1 | - | 34 | 5215 | ||
AK092007.1 | - | 1 | 1461 | |||
BC008073.2 | - | 4 | 353 | |||
BC150271.1 | - | 34 | 5052 | 9007243 | ||
D87071.1 | - | 34 | 5052 | |||
KC602455.1 | - | 45 | 6505 |