Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF144A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014746.4 | NP_055561.2 | 5 | 734 | ||
XM_005246200.3 | XP_005246257.1 | 5 | 1925 | |||
XM_005246202.4 | XP_005246259.1 | 4 | 565 | |||
XM_017005396.1 | XP_016860885.1 | 5 | 1901 | |||
XM_017005397.1 | XP_016860886.1 | 5 | 559 | |||
XM_017005398.1 | XP_016860887.1 | 5 | 672 | |||
XM_017005399.1 | XP_016860888.1 | 5 | 672 | |||
XM_017005400.1 | XP_016860889.1 | 5 | 672 | |||
XM_017005401.1 | XP_016860890.1 | 5 | 672 | |||
XM_017005402.1 | XP_016860891.1 | 5 | 672 | |||
XM_017005403.1 | XP_016860892.1 | 5 | 670 | |||
XM_017005404.1 | XP_016860893.1 | 4 | 542 | |||
GenBank mRNA | AK293854.1 | - | 5 | 671 | ||
BC050373.1 | - | 5 | 674 | 5722878 | ||
D79983.1 | - | 5 | 640 | |||
DA234807.1 | - | 5 | 553 |