Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102553208 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006254077.2 | XP_006254139.1 | 3 | 562 | ||
XM_006254078.2 | XP_006254140.1 | 3 | 715 |
Gene Symbol: LOC686142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751717.3 | XP_003751765.1 | 4 | 600 | ||
XM_006254133.2 | XP_006254195.1 | 3 | 1203 | |||
XM_006254135.2 | XP_006254197.1 | 3 | 344 |