Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267059.1 | NP_001253988.1 | 14 | 1497 | ||
NM_001267060.1 | NP_001253989.1 | 13 | 1458 | |||
NM_001267061.1 | NP_001253990.1 | 14 | 1368 | |||
NM_014748.3 | NP_055563.1 | 14 | 1533 | |||
NR_049782.1 | 14 | 1658 | ||||
NR_049783.1 | 15 | 1631 | ||||
NR_049784.1 | 15 | 1607 | ||||
NR_049785.1 | 13 | 1540 | ||||
NR_049786.1 | 14 | 1489 | ||||
NR_049787.1 | 12 | 1340 | ||||
NR_049788.1 | 11 | 1270 | ||||
XM_011533203.1 | XP_011531505.1 | 13 | 1307 | |||
XM_017005405.1 | XP_016860894.1 | 12 | 1477 | |||
GenBank mRNA | AK129689.1 | - | 14 | 1417 | ||
AK293252.1 | - | 15 | 1507 | |||
AK293464.1 | - | 14 | 1349 | |||
AK295278.1 | - | 12 | 1219 | |||
AK297054.1 | - | 14 | 1542 | |||
AK298620.1 | - | 14 | 1368 | |||
AK298856.1 | - | 14 | 1370 | |||
AK298869.1 | - | 13 | 1319 | |||
AK300144.1 | - | 14 | 1380 | |||
AK300641.1 | - | 12 | 1211 | |||
AK308487.1 | - | 13 | 1419 | |||
AK316373.1 | - | 14 | 1312 | |||
BC002524.1 | - | 14 | 1394 | 3140610 | ||
BC002610.1 | - | 14 | 1383 | 3143104 | ||
BC014620.2 | - | 14 | 1394 | 3010648 | ||
BC050590.1 | - | 14 | 1388 | 6044270 | ||
BT007167.1 | - | 14 | 1285 | |||
CR457081.1 | - | 14 | 1285 | |||
D31764.1 | - | 14 | 1507 |