Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naa16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751517.3 | XP_003751565.1 | 12 | 1514 | ||
XM_008770925.1 | XP_008769147.1 | 12 | 1514 |