Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024649.4 | NP_078925.3 | 13 | 1314 | ||
GenBank mRNA | AF503941.1 | - | 13 | 1314 | ||
AK027243.1 | - | 11 | ||||
AK095638.1 | - | 12 | 1321 | |||
AK294962.1 | - | 5 | 548 | |||
AK296311.1 | - | 9 | 1029 | |||
BC047642.1 | - | 12 | 1273 | 4824649 | ||
BC109064.2 | - | 13 | 1307 | 40033833 | ||
BC109065.1 | - | 10 | 920 | 40033834 | ||
BX647612.1 | - | 13 | 1343 |