Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CALM2 SIRNA Assays ›
Gene Symbol:
Calm2
Gene Name:
calmodulin 2
Gene Aliases:
Cam
Chromosome Location:
Chr.6: 11064750 - 11080134 on Build Rnor_6.0
UniGene ID:
Rn.5968
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s189248
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Calm2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
BC058485.1
-
280
6921326
FQ211494.1
-
280
FQ211970.1
-
276
FQ212357.1
-
277
FQ212579.1
-
276
FQ212640.1
-
276
FQ212691.1
-
276
FQ212789.1
-
280
FQ212869.1
-
280
FQ213028.1
-
276
FQ213029.1
-
274
FQ213091.1
-
276
FQ213115.1
-
282
FQ213200.1
-
279
FQ213216.1
-
276
FQ213310.1
-
169
FQ213366.1
-
275
FQ213496.1
-
279
FQ213520.1
-
280
FQ213526.1
-
276
FQ213581.1
-
276
FQ213598.1
-
276
FQ213637.1
-
276
FQ213695.1
-
276
FQ213703.1
-
276
FQ213760.1
-
276
FQ213762.1
-
281
FQ213913.1
-
276
FQ213941.1
-
280
FQ213981.1
-
277
FQ213985.1
-
170
FQ213999.1
-
275
FQ214059.1
-
280
FQ214181.1
-
280
FQ214262.1
-
280
FQ214491.1
-
276
FQ214500.1
-
276
FQ214519.1
-
276
FQ214525.1
-
276
FQ214540.1
-
280
FQ215159.1
-
280
FQ215317.1
-
276
FQ215600.1
-
280
FQ216397.1
-
276
FQ216471.1
-
277
FQ218091.1
-
276
FQ220250.1
-
276
FQ220325.1
-
276
FQ220698.1
-
280
FQ220779.1
-
276
FQ222184.1
-
279
FQ223357.1
-
280
FQ225272.1
-
276
FQ226994.1
-
280
FQ229228.1
-
276
FQ229580.1
-
280
FQ229619.1
-
276
FQ229790.1
-
276
FQ229905.1
-
275
FQ231022.1
-
280
FQ231702.1
-
280
FQ233337.1
-
279
FQ234377.1
-
280
FQ235266.1
-
277
M19312.1
-
259
Back To Top
More Information
Panther Classification:
Molecular Function -
calmodulin-related
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-slhn9:80/100.66.78.111:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline