Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CALM2 SIRNA Assays ›
Gene Symbol:
Calm2
Gene Name:
calmodulin 2
Gene Aliases:
Cam
Chromosome Location:
Chr.6: 11064750 - 11080134 on Build Rnor_6.0
UniGene ID:
Rn.5968
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s189249
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Calm2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
BC058485.1
-
157
6921326
FQ211494.1
-
157
FQ211970.1
-
153
FQ212357.1
-
154
FQ212579.1
-
153
FQ212640.1
-
153
FQ212691.1
-
153
FQ212789.1
-
157
FQ212869.1
-
157
FQ213028.1
-
153
FQ213029.1
-
151
FQ213091.1
-
153
FQ213115.1
-
159
FQ213200.1
-
156
FQ213216.1
-
153
FQ213310.1
-
46
FQ213366.1
-
152
FQ213496.1
-
156
FQ213520.1
-
157
FQ213526.1
-
153
FQ213581.1
-
153
FQ213598.1
-
153
FQ213637.1
-
153
FQ213695.1
-
153
FQ213703.1
-
153
FQ213760.1
-
153
FQ213762.1
-
158
FQ213913.1
-
153
FQ213941.1
-
157
FQ213981.1
-
154
FQ213985.1
-
47
FQ213999.1
-
152
FQ214059.1
-
157
FQ214181.1
-
157
FQ214262.1
-
157
FQ214491.1
-
153
FQ214500.1
-
153
FQ214519.1
-
153
FQ214525.1
-
153
FQ214540.1
-
157
FQ215159.1
-
157
FQ215317.1
-
153
FQ215600.1
-
157
FQ216397.1
-
153
FQ216471.1
-
154
FQ218091.1
-
153
FQ220250.1
-
153
FQ220325.1
-
153
FQ220698.1
-
157
FQ220779.1
-
153
FQ222184.1
-
156
FQ223357.1
-
157
FQ225272.1
-
153
FQ226994.1
-
157
FQ229228.1
-
153
FQ229580.1
-
157
FQ229619.1
-
153
FQ229790.1
-
153
FQ229905.1
-
152
FQ231022.1
-
157
FQ231702.1
-
157
FQ233337.1
-
156
FQ234377.1
-
157
FQ235266.1
-
154
M17069.1
-
155
M19312.1
-
136
Back To Top
More Information
Panther Classification:
Molecular Function -
calmodulin-related
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-x88h5:80/100.66.78.96:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline