Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014763.3 | NP_055578.2 | 2, 3 | 251 | ||
GenBank mRNA | AK291250.1 | - | 2, 3 | 237 | ||
AK295578.1 | - | 2, 3 | 223 | |||
BC030144.2 | - | 2, 3 | 236 | 4763405 | ||
BI551514.1 | - | 2, 3 | 256 | |||
BQ432684.1 | - | 2, 3 | 254 | |||
D14660.1 | - | 2, 3 | 224 |