Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014763.3 | NP_055578.2 | 6 | 846 | ||
GenBank mRNA | AK291250.1 | - | 6 | 832 | ||
AK295578.1 | - | 6 | 818 | |||
BC030144.2 | - | 6 | 831 | 4763405 | ||
D14660.1 | - | 6 | 819 |