Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IP6K1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006115.2 | NP_001006115.1 | 5 | 1226 | ||
NM_001242829.1 | NP_001229758.1 | 6 | 1915 | |||
NM_153273.3 | NP_695005.1 | 6 | 1577 | |||
GenBank mRNA | AK289772.1 | - | 5 | 1382 | ||
AK295689.1 | - | 5 | 1102 | |||
BC012944.1 | - | 5 | 1378 | 3872311 | ||
D87452.2 | - | 6 | 1564 |