Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calml4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127575.1 | NP_001121047.1 | 4 | 391 | ||
XM_006243243.2 | XP_006243305.1 | 3 | 341 | |||
GenBank mRNA | BC158759.1 | - | 4 | 343 | 7099908 |